Skip to Content
Merck
All Photos(1)

Key Documents

UC0E04

Sigma-Aldrich

UCOE® Dual Expression Hygromycin Vector Set

Synonym(s):

UCOE Mammalian Protein Expression Vectors, UCOE Mammalian Protein Expression Plasmids, Ubiquitous Chromatin Opening Element, UCOE04

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€1,310.00

€1,310.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
1 KIT
€1,310.00

About This Item

UNSPSC Code:
41106621
eCl@ss:
34360190
NACRES:
NA.51

€1,310.00


Estimated to ship on22 April 2025Details


shipped in

dry ice

Quality Level

General description

The Ubiquitous Chromatin Opening Element (UCOE) technology offers significant advances in recombinant mammalian protein expression. Unlike traditional vectors, UCOE-containing vectors have the capacity to ease the process of isolating cell line clones that express high-levels of recombinant proteins. UCOE sequences promote reproducible and stable high level expression of a linked gene by altering chromatin structure to a transcriptionally permissive state that negates epigenetic-mediated (e.g. DNA methylation) silencing.

CET 1019 HD-hygro-SceI is a mammalian expression plasmid vector containing both the mouse Rps3 UCOE and the human HNRPA2B1-CBX3 UCOE each upstream of a strong human cytomegalovirus (hCMV) immediate early promoter-enhancer element.

CET 1019 AD-hygro-SceI is a mammalian expression plasmid vector containing both the mouse Rps3 UCOE and the human HNRPA2B1-CBX3 UCOE each upstream of a guinea pig CMV (gpCMV) promoter-enhancer element.

The CET 1019 HD-hygro-SceI and CET 1019 AD-hygro-SceI vectors possess the following features:

  • A multiple cloning site downstream of the UCOE-CMV combinations containing unique restriction enzyme sites to permit easy insertion of any gene of interest (cDNA or genomic)
  • A polyadenylation element from the SV40 early region located downstream of the multiple cloning site for appropriate mRNA 3 end formation
  • A hygromycin antibiotic resistance gene for selection of stably transfected mammalian cells
  • A β-lactamase gene for plasmid selection and propagation in transformed prokaryotic cells in the presence of ampicillin
  • An I-SceI homing restriction endonuclease site for linearization of constructs prior to transfection into mammalian cells, which favors integration into the target cell genome via the pre-generated free DNA ends and thus retains the integrity of the UCOE-CMV-transgene cassettes


THIS PRODUCT IS ONLY AVAILABLE FOR SALE TO ACADEMIC INSTITUTIONS OR NOT-FOR-PROFIT ENTITIES FOR USE UNDER THIS PRODUCT LABEL LICENSE. FOR INFORMATION ON COMMERCIAL LICENSING OF THE PATENTED UCOE TECHNOLOGY, PLEASE CONTACT [email protected].

Application

Research Category
General Cell Biology

All
Research Sub Category
All
Ubiquitous Chromatin Opening Element (UCOE) technology offers significant advances in recombinant mammalian protein expression by removing epigenetic related gene silencing.

Components

1) 10ug CET 1019 HD-hygro-SceI (+) Vector (CS221307)

2) 10ug CET 1019 AD-hygro-SceI (+) Vector (CS221297)

3) 10ug DC HD-hygro (-) Vector (CS221298)

4) 10ug DC AD-hygro (-) Vector (CS221299)

Storage and Stability

Plasmids should be stored at -20C. Avoid repeated freeze-thaw events.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

UCOE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Antoniou et al.
Genomics, 82(3), 269-279 (2003-08-09)
The genetic elements that are responsible for establishing a transcriptionally competent, open chromatin structure at a region of the genome that consists only of ubiquitously expressed, housekeeping genes are currently unknown. We demonstrate for the first time through functional analysis
Fang Zhang et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 18(9), 1640-1649 (2010-07-01)
DNA methylation may restrict the activity of gene transfer vectors due to inadvertent silencing. In P19 embryonic carcinoma cells in vitro, we found that transgene expression regulated by the SFFV LTR and EF1 alpha promoter declined rapidly within 16 days

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service