Key Documents
SYMBA4RP2
Millipore
Steritest® Symbio Advanced Requalification Protocol
A4 format
Select a Size
1 EA
€776.00
€776.00
Estimated to ship on22 April 2025Details
Request more information
Select a Size
Change View
1 EA
€776.00
About This Item
UNSPSC Code:
41116500
NACRES:
NB.79
€776.00
Estimated to ship on22 April 2025Details
Request more information
Quality Level
compatibility
for use with Steritest® Symbio pump.
General description
Steritest® Symbio Advanced Requalification protocol is designed as a ready to use guide to perform the requalification of the Steritest® Symbio pump. The requalification protocol can be used to re-certify consistent performance of the pumps upon encountering any change or at regular intervals. On top of some IQ and OQ tests it also covers the periodic review of operAtor training, SOPs, and OOS results.
Application
Steritest® Symbio Advanced Requalification protocol is used for the re-qualification of Steritest® Symbio pump which is used to perform sterility testing for pharmaceutical products.
Legal Information
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany