Skip to Content
Merck
All Photos(2)

Key Documents

SSWP04700

Millipore

MCE Membrane Filter, 3.0 μm Pore Size

MCE Membrane Filter, 3.0 μm Pore Size
1 of 1 reviewers received a sample product or took part in a promotion

MF-Millipore, filter diam. 47 mm, hydrophilic

Synonym(s):

Hydrophilic MCE membrane filter cut discs, MF-Millipore Membrane Filter, 3 µm pore size, Mixed Cellulose Ester membrane filter discs

Sign Into View Organizational & Contract Pricing

Select a Size

100 FILTERS
€213.00

€213.00


Estimated to ship on04 April 2025Details



Select a Size

Change View
100 FILTERS
€213.00

About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

€213.00


Estimated to ship on04 April 2025Details


material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

30 L/min-cm2 air flow rate
320 mL/min-cm2 water flow rate
75 °C max. temp.

diam.

47 mm

filter diam.

47 mm

thickness

150 μm

gravimetric extractables

6%

refractive index

n/D 1.495

matrix

MF-Millipore

pore size

3.0 μm pore size
83 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥0.7 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: SSWP

Application

  • QC of fluid holding tanks
  • Fluid monitoring
  • Air monitoring
  • Particle collection
  • Particle analysis

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service