SK1M004W01
Pellicon® XL50 Cassette with Biomax® Membrane
NMWCO 5 kDa
Synonym(s):
Pellicon® XL50 with Biomax 5 kDa Membrane, C Screen, 50 cm2
About This Item
material
polyethersulfone
polypropylene
polypropylene screen
feature
holdup volume 3.2 mL
manufacturer/tradename
Pellicon®
parameter
0.03-0.05 L/min recirculation rate
2.8 bar max. transmembrane pressure (40 psig)
5.5 bar max. inlet pressure (80 psi) at 50 °C
technique(s)
analytical sample preparation: suitable
protein purification: suitable
L
18.8 cm (7.4 in.)
W
3 cm (1.2 in.)
membrane nominal area
50 cm2
gravimetric extractables
≤7.5 mg/device
matrix
Biomax®
pore size
5 kDa NMWCO
pH
1-14(cleaning)
1-14(continuous)
General description
Features and Benefits
Packaging
Other Notes
Analysis Note
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service