Skip to Content
Merck
All Photos(1)

Key Documents

SJHVM4710

Millipore

Millicup-HV, 0.45 µm, PVDF, non-sterile

Non sterile Universal bottletop vacuum filter with Hydrophilic PVDF Durapore membrane.

Synonym(s):

0.45 µm Millicup-HV, PVDF Millicup

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€192.00

€192.00


Available to ship on14 April 2025Details



Select a Size

Change View
10 EA
€192.00

About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.24

€192.00


Available to ship on14 April 2025Details


material

PVDF membrane
high-density polyethylene
high-density polyethylene housing
polyethylene housing

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Millicup

parameter

27 inHg vacuum limit (685 mmHg) at 25 °C (differential)
3 L sample volume
50 °C max. temp.

technique(s)

vacuum filtration: suitable

H

120 mm

diam.

8.4 cm

filter diam.

47 mm

filtration area

11.5 cm2

funnel capacity

300 mL

matrix

Durapore®

pore size

0.45 μm pore size

fitting

inlet funnel
outlet bottle top (Universal)

application(s)

filtration

shipped in

ambient

General description

The Millicup Filter Unit provides fast and effective filtration of solvents and buffers (up to 3L) used in HPLC and other analytical techniques.

Features & Benefits:
•No preparation or clean-up saves time
•Available with low-protein binding hydrophilic Durapore PVDF, hydrophobic Fluoropore PTFE, and hydrophilic LCR PTFE membranes
•Ultraclean design and safe operation

Applications:
Mobile Phase Preparation, Buffer Filtration

Application

Direct patient care: No

Principle

Vacuum

Physical form

Color Code: Natural

Legal Information

Durapore is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service