SCT150
Firefly Luciferase HTS Assay
Glow-type luciferase assay for the quantification of firefly luciferase gene expression in mammalian cells with signal half-life of about 3 hours
Synonym(s):
Firefly Luciferase Assay, Bioluminescent Assay
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
cell based assay: suitable
detection method
chemiluminescent
shipped in
ambient
Related Categories
General description
Firefly luciferase is widely used as a reporter for studying gene regulation and function, and for pharmaceutical screening. It is a very sensitive genetic reporter due to the absence of endogenous luciferase activity in mammalian cells or tissues. Firefly luciferase is a 62,000 Dalton protein, which is active as a monomer and does not require subsequent processing for its activity. The enzyme catalyzes ATP-dependent D-luciferin oxidation to oxyluciferin, producing light emission centered at 560 nm. Light emitted from the reaction is directly proportional to the number of luciferase enzyme molecules.
However, the light production resulting from the reaction leads to formation of suicidal adenyl-oxyluciferin at the enzyme surface. It results in very short half-life of the light emission with a flash-type kinetics. The Firefly Luciferase HTS Assay is designed for is a proprietary mixture of substances that modify the enzymatic reaction to produce a long-lasting signal (steady-glow) by preventing the formation of adenyl-oxyluciferin at the enzyme surface. It is a homogeneous high sensitivity firefly luciferase reporter gene assay kit for the quantification of firefly luciferase expression in mammalian cells with signal half-life of about 3 hours (Figure 2). Glow-type luciferase assays have lower luminescence signal compared to flash-type assays. The sensitivity and limit of detection of the assay will depend on luciferase expression levels in your experimental system as well as luminometer sensitivity.
However, the light production resulting from the reaction leads to formation of suicidal adenyl-oxyluciferin at the enzyme surface. It results in very short half-life of the light emission with a flash-type kinetics. The Firefly Luciferase HTS Assay is designed for is a proprietary mixture of substances that modify the enzymatic reaction to produce a long-lasting signal (steady-glow) by preventing the formation of adenyl-oxyluciferin at the enzyme surface. It is a homogeneous high sensitivity firefly luciferase reporter gene assay kit for the quantification of firefly luciferase expression in mammalian cells with signal half-life of about 3 hours (Figure 2). Glow-type luciferase assays have lower luminescence signal compared to flash-type assays. The sensitivity and limit of detection of the assay will depend on luciferase expression levels in your experimental system as well as luminometer sensitivity.
Application
Firefly Luciferase Assay
Glow-type luciferase assay for the quantification of firefly luciferase gene expression in mammalian cells with signal half-life of about 3 hours
Research Category
Cell Signaling
Cell Signaling
Research Sub Category
Live Cell Dye
Live Cell Dye
Components
1) Assay Buffer (CS224524): 1X100 ml
2) D-Luciferin (CS224523): 1 X 25 mg
2) D-Luciferin (CS224523): 1 X 25 mg
Physical form
Liquid
Storage and Stability
Store Firefly Luciferase HTS Assay at -80°C. Kit components are stable for at least six months from date of receipt when stored as recommended. Avoid repeated freeze-thaw cycles.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Firefly luciferase is a sensitive reporter for gene studies due to its absence in mammalian cells or tissues.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service