The SCT125 is neither a pre-stain nor a loading dye. According to the data sheet, the recommendation is to first run the gel, and then immerse it in a 1 to 10,000 dilution of SCT125 post-electrophoresis. Instructions for its use can be found on the product's data sheet, found in the DOCUMENTATION section of the Product Detail Page: https://www.sigmaaldrich.com/product/mm/sct125#product-documentation
SCT125
GelGreen® Nucleic Acid Stain 10000X Water
GelGreen is a fluorescent nucleic acid stain designed to replace the highly toxic ethidium bromide (EtBr) for staining dsDNA, ssDNA or RNA in agarose gels or polyacrylamide gels.
Synonym(s):
Fluorescent DNA gel stain
About This Item
Recommended Products
form
liquid
technique(s)
PCR: suitable
solubility
water: soluble
suitability
suitable for nucleic acid staining
detection method
fluorometric
Related Categories
General description
The dye is noncytotoxic and non-mutagenic at concentrations well above the working concentrations used in gel staining. GelGreen successfully passed environmental safety tests in compliance with CCR Title 22 Hazardous Waste Characterization, under which GelGreen is not classified as hazardous waste.
Spectral Properties
Absorbance: Standard Transilluminator (254 nm) such as a Dark Reader
Emission: Long path green filter such as a SYBR filter or GelStar filter.
Application
Cell Culture
Live Cell Dye
Quality
Absorbance: Standard Transilluminator (254 nm) such as a Dark Reader
Emission: Long path green filter such as a SYBR filter or GelStar filter.
Physical form
Storage and Stability
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
188.6 °F - (Dimethylsulfoxide)
Flash Point(C)
87 °C - (Dimethylsulfoxide)
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Is SCT125 used as a pre-stain mixed into the gel prior to the electrophoresis run, or is it a loading dye that should be added to the samples immediately before being loaded into the wells?
Is SCT125 used as a pre-stain mixed into the gel prior to the electrophoresis run, or is it a loading dye that should be added to the samples immediately before being loaded into the wells?
1 answer-
Helpful?
-
-
Would gel green work for a gel reader that uses blue light instead of UV light? The excitation needs to be about 480 nm
1 answer-
Yes, this product compatible with either a 254 nm UV transilluminator or a gel reader equipped with visible light excitation (such as a 488 nm laser-based gel scanner or a Dark Reader).
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service