Skip to Content
Merck
All Photos(1)

Key Documents

SCR067

Sigma-Aldrich

Human Mesenchymal Stem Cell Characterization Kit

The Human Mesenchymal Stem Cell Characterization Kit contains a panel of positive & negative selection markers for the characterization of the mesenchymal stem cell population in human samples.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.75
Pricing and availability is not currently available.

Quality Level

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable
immunocytochemistry: suitable

input

sample type mesenchymal stem cell(s)

shipped in

dry ice

General description

The Human Mesenchymal Stem Cell Characterization Kit (Catalog Number SCR067) contains a panel of positive and negative selection markers for the characterization of the mesenchymal stem cell population in human samples Positive cell markers include antibodies directed against cell-surface molecules present on mesenchymal stem cells: CD44, CD90, STRO-1, and CD146. In addition, two specific hematopoeitic cell surface markers are provided whose expressions should not be present on mesenchymal stem cells: CD14 (present on leukocytes) and CD19 (present on B-lymphocytes).

Components

Mouse anti-HCAM

Mouse anti-THY-1 (CD90)

Mouse anti-STRO-1

Mouse anti-MCAM (CD146)

Mouse anti-CD19

Mouse anti-CD14

Storage and Stability

When stored at the recommended storage conditions (refer to Kit Components), components are stable up to the expiration date. Do not expose to elevated temperatures. Discard any remaining reagents after the expiration date.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and characterization of human periodontal ligament (PDL) stem cells (PDLSCs) from the inflamed PDL tissue: in vitro and in vivo evaluations.
Jung-Chul Park,Jeong-Min Kim,Im-Hee Jung,Jane C Kim,Seong-Ho Choi,Kyoo-Sung Cho,Chang-Sung Kim
Journal of Clinical Periodontology null
Khondoker M Akram et al.
Respiratory research, 14, 9-9 (2013-01-29)
Mesenchymal stem cells (MSC) are in clinical trials for widespread indications including musculoskeletal, neurological, cardiac and haematological disorders. Furthermore, MSC can ameliorate pulmonary fibrosis in animal models although mechanisms of action remain unclear. One emerging concept is that MSCs may
Acquisition of human alveolar bone-derived stromal cells using minimally irrigated implant osteotomy: in vitro and in vivo evaluations.
Jung-Chul Park,Jane C Kim,Yong-Tae Kim,Seong-Ho Choi,Kyoo-Sung Cho,Gun-Il Im et al.
Journal of Clinical Periodontology null
The dynamic healing profile of human periodontal ligament stem cells: histological and immunohistochemical analysis using an ectopic transplantation model.
Y-T Kim,J-C Park,S-H Choi,K-S Cho,G-I Im,B-S Kim,C-S Kim
Journal of Periodontal Research null
Anna Badner et al.
Journal of neuroinflammation, 15(1), 219-219 (2018-08-05)
The spleen plays an important role in erythrocyte turnover, adaptive immunity, antibody production, and the mobilization of monocytes/macrophages (Mφ) following tissue injury. In response to trauma, the spleen initiates production of inflammatory cytokines, which in turn recruit immune cells to

Articles

Highly characterized human mesenchymal stem cell lines from bone-marrow, adipose and umbilical cord along with serum-free MSC expansion and differentiation media.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service