Skip to Content
Merck
All Photos(1)

Key Documents

SCR047

Sigma-Aldrich

Pancreatic Cell DTZ Detection Assay

The Pancreatic Cell DTZ Detection Assay provides researcher a simple & quick method to identify insulin-producing beta cells from a mixed cell culture preparation or from pancreatic tissues.

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€308.00

€308.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€308.00

About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.84

€308.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®
Millipore

technique(s)

cell culture | stem cell: suitable

input

sample type pancreatic stem cell(s)

detection method

colorimetric

shipped in

dry ice

General description

The Pancreatic Cell DTZ Detection Assay provides researcher a simple and quick method to identify insulin-producing beta cells from a mixed cell culture preparation or from pancreatic tissues The kit contains pre-mixed Dithizone Stain, Rinsing Solution and filters for fast preparation and optimized results. Millipore′s Pancreatic Cell DTZ Detection Asssay (SCR047) takes advantage of the high zinc content of pancreatic beta cells and the known action of dithizone, a zinc-chelating agent, to provide a quick and simple enzymatic reaction that allows researcher to reliably identify insulin-producing beta cells from a mixed cell culture preparation or from pancreatic tissues. Along with selectively staining live pancreatic beta-islet cells crimson red, the enzymatic reaction has the advantage of being completely reversible. Kit components include staining and rinse solutions along with filters required for the live staining reactions. Sufficient reagents are supplied to provide for 10 enzymatic reactions at 10 mL volume.
The islets of Langerhans are groups of specialized cells that are dispersed throughout the pancreas. They produce a number of hormones including glucagon, insulin, Pancreatic polypeptide and somatostatin. Insulin is the most critical of these, as it helps regulate glucose metabolism. Insulin is synthesized from a precursor molecule, proinsulin, in the beta cells of the islets of Langerhans. The conversion of proinsulin into the active hormone, insulin, is accomplished by proteolytic enzymes known as prohormone convertases (PC1 and PC2) as well as the exoprotease carboxypeptidase E. These proteolytic enzymes remove the center portion of the molecule, or C-peptide, and the remaining peptide chains (A and B) are cross-linked by disulfide bonds. The crosslinked peptides are the insulin monomer. As the concentration of the protein increases, the monomers assemble into dimers and in the presence of zinc, into hexameric crystals characterized by two Zn2+ atoms per hexameric unit. It is this 2-Zn insulin hexamer that represents the active conformation of the secreted hormone. Given its prominent role in the synthesis, storage and secretion of insulin, it is not surprising that zinc is highly expressed in pancreatic beta cells.

Application

Research Category
Stem Cell Research
The Pancreatic Cell DTZ Detection Assay provides researcher a simple & quick method to identify insulin-producing beta cells from a mixed cell culture preparation or from pancreatic tissues.

Components

1 mL of 100X DTZ Stain Solution (Part No: 2006182)

2 - 150 mL of 1X DTZ Rinse Solution (Part No: 2006183)

10 Millex GP 0.22 uM filters (Part No: SLGPR33RB)

12 Plastic Syringes, 20 mL w/ Luer slip (Part No: XX1102012)

Storage and Stability

Store DTZ Stain at -20C. Store all other reagents at room temperature.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the expiry date of this product?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service