This product is not scheduled to be or listed as a GMP grade or clinical grade product. This product falls under a Quality Level 100, products with a quality level 100 are not suitable for GMP use.
SCM141
PLTMax Human Platelet Lysate
PLTMax Human Platelet Lysate is a growth factor rich supplement that is a superior alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) culture. 100ml.
Synonym(s):
hPL, hPL Human Platelet Lysate Supplement
Select a Size
€449.00
Select a Size
About This Item
€449.00
Recommended Products
biological source
human
Quality Level
form
liquid
manufacturer/tradename
Chemicon®
technique(s)
cell culture | stem cell: suitable
input
sample type mesenchymal stem cell(s)
shipped in
dry ice
General description
PLTMax Human Platelet Lysate is derived from normal human donor platelets collected at U.S. blood centers. Multiple donors units are pooled in large batch sizes and manufactured to produce a consistent product. Heparin (porcine) approved for human intravenous use is used in the preparation of PLTMax Human Platelet Lysate. This product contains human derived materials and must be treated as potentially infectious. Universal precautions for biological samples should be used in handling and disposal.
Application
• Add Heparin to the media at a final concentration of 2 U/mL. Failure to add heparin may result in coagulation of certain media.
• For optimal growth of human mesenchymal stem cells, add 5% vol/vol PLTMax Human Platelet Lysate to usual cell culture medium such as DMEM and αMEM.
• Plate cells at approximately 1.5x103 cells per cm2.
• Do not allow primary MSC confluence to exceed 80%.
Particulate Formation
• Product has been aseptically filtered with 0.2 µm filters. Particulates will be observed after thawing. Particulate formation is normal will not affect performance even in the presence of large particulates. Filtration is not recommended.
• To reduce particulates from forming, minimize freeze/thaw cycles and extended storage at 4°C. Some particulate formation can be reversed with brief incubation at 37°C, if desired.
Stem Cell Research
Mesenchymal Stem Cells
Quality
Each lot is extensively tested for the following parameters:
pH: 6.8-7.8
Total protein: 4.0-6.5 g/dl
Endotoxin USP <8>: <10 EU/mL
Mycoplasma: Not detected
Sterility Testing: Negative for growth
Storage and Stability
Legal Information
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Highly characterized human mesenchymal stem cell lines from bone-marrow, adipose and umbilical cord along with serum-free MSC expansion and differentiation media.
Read types of stem cells including multipotent stem cells, Pluripotent stem cells and iPSCs and their applications in basic stem cell research, stem cell therapy and disease modelling
Development of a novel serum-free and xeno-free human mesenchymal stem cell (MSC) osteocyte differentiation media.
PLTMax® Human Platelet Lysate (hPL) is a superior serum-free and xeno-free media supplement alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) cultures.
Protocols
Information about mesenchyme, specifically mesenchymal stem cell procotols. Step-by-step cell culture protocols for mesenchymal stem cell (MSC) isolation, expansion and differentiation.
-
GMP/CLINICAL GRADE로 개발이 계획되어 있는지 문의드립니다.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service