Skip to Content
Merck
All Photos(1)

Key Documents

SCM102

Sigma-Aldrich

Cardiomyocyte Differentiation Medium

The Cardiomyocyte Differentiation Medium is available in a 500 mL format & has been optimized & validated for Cell Differentiation.

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€153.00

€153.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 ML
€153.00

About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

€153.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Quality Level

form

liquid

technique(s)

cell differentiation: suitable

input

sample type cardiac stem cell(s)

shipped in

dry ice

General description

The Cardiomyocyte Differentiation Medium has been optimized and qualified for the preferential differentiation of freshly isolated cardiac stem cells derived from rodent origins to cardiomyocytes. Cardiac stem cells maintained in Cardiomyocyte Differentiation Medium for 12-15 days should differentiate into cardiomyocyte and express the appropriate markers, troponin I, desmin, and actinin.

Quality

Each lot of medium is tested for the optimal differentiation of cardiac stem cells to cardiomyocytes. Sterility Testing: Negative

Physical form

Cardiomyocyte Differentiation Medium is a proprietary formulation that contains fetal bovine serum.

Storage and Stability

For long term storage, maintain the media at -20°C. Prior to initial use, thaw frozen media at 4°C overnight or until it has become completely equilibrated. Maintain thawed media at 2-8°C in the dark for up to one month.

Legal Information

Accutase is a registered trademark of Innovative Cell Technologies, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service