Skip to Content
Merck
All Photos(8)

Key Documents

SCC416

Sigma-Aldrich

Py2T Mouse Mammary Tumor Cell Line

Mouse

Synonym(s):

Py2T cell line;Mouse mammary tumor cell line;Py2T Murine breast cancer cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Product SCC416 is not currently sold in your country. Contact Technical Service

Product Name

Py2T Mouse Mammary Tumor Cell Line,

biological source

mouse

Quality Level

packaging

vial of ≥1X10⁶ cells

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

−196°C

General description

≥1x106 viable Py2T Mouse Mammary Tumor Cells: (Catalog No. SCC416). Store in liquid nitrogen.

Application

  • Each vial contains > 1X106 viable cells.
  • Cells are tested negative for infectious diseases by a Mouse Essential CLEAR Panel by Charles River Animal Diagnostic Services.
  • Cells are verified to be of mouse origin and negative for interspecies contamination from human, rat, Chinese hamster, Golden Syrian hamster, and nonhuman primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination.

Py2T is a stable murine breast cancer cell line derived from the mammary tumor of MMTV-PyMT (mouse mammary tumor virus-polyoma middle tumor-antigen) transgenic mouse. PyMT is an oncogene which transforms cells in culture and leads to formation of mammary tumors in MMTV-PyMT mice by interacting with non receptor tyrosine kinase, c-Src.
These cells have a uniform cobblestone-like morphology, typical of differentiated epithelial cells, and express estrogen receptor (ER), and both basal (CK14) and luminal (CK8/18) markers. When these cells are implanted orthotopically in syngeneic FVB/N mice, they form non-metastatic invasive tumors with low or absent E-cadherin.
Epithelial-mesenchymal transition (EMT) is a process involved in the initiation of the invasion-metastasis cascade of cancer cells. 90% of breast cancer-related deaths are ascribed to systemic dissemination of cancer cells leading to their metastatic outgrowth in distant organs.


Source
The Py2T cell line was derived from a mammary tumor of a MMTV-PyMT transgenic mouse.

References
1.PLoS One 2012; 7(11):e48651. PMID: 23144919.
2.Oncogene 2015; 34(32):4190-4198. PMID: 25362852.
3.Sci Rep 2018; 8(1):12123. PMID: 30108334.
4.Ayse NK. University of Basel 2017; Dissertation. https://edoc.unibas.ch/59030/1/Ph.D.%20Dissertation.pdf

Features and Benefits

Py2T undergo epithelial-mesenchymal transition (EMT) in response to TGFβ in vitro. Long term treatment with TGFβ leads to formation of a mesenchymal subline of cells (Py2T LT) with elongated morphology expressing mesenchymal markers such as vimentin, fibronectin, and N-cadherin. Orthotopic implantation of Py2T LT cells has been reported to form microscopic lung metastatic lesions in mice. These cell lines will serve as a versatile model to investigate the molecular mechanisms underlying EMT.

Storage and Stability

Py2T cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages without significantly affecting cell marker expression and function.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Restricted Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service