Skip to Content
Merck
All Photos(1)

Key Documents

SCC162

Sigma-Aldrich

CFTE29o- Human Cystic Fibrosis Tracheal Epithelial Cell Line

Human

Synonym(s):

CFTE29o, CFTE-29o-

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81
Product SCC162 is not currently sold in your country. Contact Technical Service

Product Name

CFTE29o- Human Cystic Fibrosis Tracheal Epithelial Cell Line, CFTE29o- human cystic fibrosis tracheal epithelial cell line is a useful model for cystic fibrosis research and for studying chloride ion transport and ion transport in human airways.

biological source

human

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable

shipped in

ambient

General description

Cystic Fibrosis (CF) is a lethal autosomal recessive disease caused by mutations in the CF transmembrane conductance regulator (CFTR) gene which functions as a cAMP-activated and phosphorylated-regulated Cl channel. The predominant mutation in the CFTR gene is a trinucleotide deletion that results in loss of a phenylalanine at amino acids 508 (ΔF508) in the CFTR protein. This mutation accounts for ~66% of all CF alleles [1].

CFTE29o- is a human CF tracheal epithelial cell line, derived from a 21-yr old male CF patient homozygous for the ΔF508 CFTR mutation and immortalized with the origin-of-replication defective SV40 plasmid (pSVori-) [1]. The cell line express keratin, indicating an epithelial cell origin, along with a calcium-dependent cell adhesion protein, cellCAM 120/80 [1], and ZO-1 indicating the ability to form tight junctions. CFTE29o- displays ion transport properties characteristic of cystic fibrosis such as defective cAMP-dependent chloride transport. Upon treatment with the calcium ionophore, ionomycin, CFTE29o- secretes chloride, albeit at reduced levels relative to normal cells. The cells express CFTR mRNA, and small amounts of CFTR protein as assessed by Western blot hybridization [1]. Karyotype analysis indicates that 70% of the cells contain two copies of chromosome 7 [1].

CFTE29o- is a useful model for cystic fibrosis and for studying chloride ion transport and ion transport in human airways. The cells are useful for testing candidate chemotherapeutic drugs for their potential as treatments for ameliorating CF pathologies.

Cell Line Description

Epithelial Cells

Application

CFTE29o- human cystic fibrosis tracheal epithelial cell line is a useful model for cystic fibrosis research and for studying chloride ion transport and ion transport in human airways.
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Quality

• Each vial contains ≥ 1X10⁶ viable cells.
• Cells are tested by PCR and are negative for HPV-16, HPV-18, Hepatitis A, C, and HIV-1 & 2 viruses as assessed by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are negative for mycoplasma contamination.
• Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Kunzelmann et al.
American journal of respiratory cell and molecular biology, 8(5), 522-529 (1993-05-01)
The development of transformed human airway epithelial cell lines has been important in advancing the understanding of the biochemical and genetic mechanisms underlying the cystic fibrosis (CF) defect. Since the most common mutation associated with CF is a phenylalanine deletion

Articles

16HBE14o- human bronchial epithelial cells used to model respiratory epithelium for the research of cystic fibrosis, viral pulmonary pathology (SARS-CoV), asthma, COPD, effects of smoking and air pollution. See over 5k publications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service