Skip to Content
Merck
All Photos(1)

Documents

SCC057

Sigma-Aldrich

FibroGRO® Inactivated Xeno-Free Human Foreskin Fibroblasts

The FibroGROTM Xeno-Free Human Foreskin Fibroblasts (Mitomycin C treated) are derived from normal human foreskin (male neonate) & have been isolated & manufactured under xeno-free conditions.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.71

biological source

human foreskin fibroblasts

Quality Level

packaging

pkg of 1 kit

manufacturer/tradename

FibroGRO®
Millipore Chemicon®

morphology

(fibroblast)

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

shipped in

ambient

General description

Millipore’s FibroGROTM Xeno-Free Human Foreskin Fibroblasts (Mitomycin C treated) are derived from normal human foreskin (male neonate) and have been isolated and manufactured under xeno-free conditions. Cells have been mitotically inactivated with Mitomycin C and validated not to replicate. Cells are supplied at a low passage number (p6 – p8), tested negative for mycoplasma and can be used as a feeder layer to maintain human embryonic stem cell culture in an undifferentiated state.
Product Source: Human foreskin fibroblasts, derived under xeno-free conditions

Cell Line Origin

Foreskin

Cell Line Description

Fibroblast Cells

Application

Research Category
Stem Cell Research
The FibroGROTM Xeno-Free Human Foreskin Fibroblasts (Mitomycin C treated) are derived from normal human foreskin (male neonate) & have been isolated & manufactured under xeno-free conditions.

Components

4 vials of Mit-C treated fibroblasts

Quality

Negative for mycoplasma, HTLV-1&2, Hepatitis B&C, HIV, Treponema palladium.

Storage and Stability

Store in Liquid Nitrogen

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
FIBROGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service