Skip to Content
Merck
All Photos(1)

Key Documents

SAMP2LID

Millipore

Samplicity® G2 Filtration System Replacement Lid

To be used with the Samplicity G2 Filtration System. Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€158.00

€158.00


Estimated to ship on08 July 2025Details



Select a Size

Change View
1 EA
€158.00

About This Item

UNSPSC Code:
41104922
eCl@ss:
32031690
NACRES:
NB.24

€158.00


Estimated to ship on08 July 2025Details


Quality Level

sterility

non-sterile

manufacturer/tradename

Samplicity®

technique(s)

HPLC: suitable
analytical sample preparation: suitable

shipped in

ambient

General description

Overview of Samplicity® G2 System

• Samplicity G2 Filtration System enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.

• Filtration for HPLC sample prep without syringes. Sample is added to an adapter funnel on a standard Millex 33 mm filter (Millex Filters for the Samplicity G2 System) or an integrated funnel and filter (Millex Samplicity filter)

• Samplicity G2 System has lower ergonomic impact than manual syringe filtration



Applications:

HPLC; Filtration of viscous or particulate laden samples for HPLC

Legal Information

SAMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrophilic PTFE Millex® syringe filters ensure accurate dissolution testing and sample preparation for HPLC analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service