Skip to Content
Merck
All Photos(1)

Key Documents

S7707FR

Sigma-Aldrich

TRAPEZE XL Telomerase Detection Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€481.00

€481.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
1 KIT
€481.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

€481.00


Estimated to ship on22 April 2025Details


Legal Information

TRAPEZE is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service