Key Documents
RPSTA4VP1
Millipore
Rapid Sterility Testing Validation Protocol
A4 Format, Milliflex® Rapid PQ Protocol
Select a Size
1 EACH
€1,450.00
€1,450.00
Available to ship on11 April 2025Details
Request more information
Select a Size
Change View
1 EACH
€1,450.00
About This Item
UNSPSC Code:
41116500
NACRES:
NA.85
€1,450.00
Available to ship on11 April 2025Details
Request more information
Quality Level
compatibility
for use with Milliflex® Rapid Systems.
General description
Rapid Sterility Testing Validation Protocol is designed as a ready to use step by step guide to perform PQ activities of rapid sterility testing using Milliflex® Rapid system. The PQ protocol will enable the QC/QA Labs to initiate the performance qualification after the IQ andOQ have been completed following the respective separate protocol. The guidelines are GMP compliant and follow EP, USP and JP regulatory guidelines.
Application
Rapid Sterility Testing Validation Protocol is used to validate rapid sterility testing procedures with ease and consistent performance while following regulatory guidelines.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany