Skip to Content
Merck
All Photos(2)

Key Documents

RMHE-120K

Millipore

MILLIPLEX® Rat Metabolic Hormone Expanded Panel - Metabolism Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

To order a Milliplex® kit, please search for your analyte of interest.

usage

 kit sufficient for 96 wells

Quality Level

species reactivity

rat

manufacturer/tradename

Milliplex®

assay range

standard curve range: 1.4-1,000 pg/mL
(TNFα)

standard curve range: 110-80,000 pg/mL
(IL-6)

standard curve range: 110-80,000 pg/mL
(Insulin)

standard curve range: 2.7-2,000 pg/mL
(Glucagon)

standard curve range: 27-20,000 pg/mL
(GLP-1 (active))

standard curve range: 27-20,000 pg/mL
(GLP-1 total)

standard curve range: 4.1-3,000 pg/mL
(GIP (total))

standard curve range: 4.1-3,000 pg/mL
(Ghrelin (active))

standard curve range: 41-30,000 pg/mL
(Amylin (active))

standard curve range: 48-35,000 pg/mL
(Leptin)

standard curve range: 6.9-5,000 pg/mL
(PYY total)

standard curve range: 6.9-5,000 pg/mL
(Pancreatic Polypeptide, PP)

standard curve range: 6.9-5,000 pg/mL
(Secretin)

standard curve range: 69-50,000 pg/mL
(C-Peptide 2)

standard curve range: 69-50,000 pg/mL
(MCP-1/CCL2)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

General description

MILLIPLEX® Rat Metabolic Hormone Expanded Panel is a 14-plex kit to be used for the simultaneous quantification of any or all of the following analytes in tissue/lysate and culture supernatant samples and rat serum or plasma: Active Amylin, C-peptide 2, Active Ghrelin, Gastric Inhibitory Polypeptide (GIP), Glucagon-like peptide-1 (GLP-1) Active or Total, Glucagon, Interleukin-6 (IL-6), Insulin, Leptin, Monocyte Chemoattractant Protein-1 (MCP-1), Pancreatic Polypeptide (PP), Peptide YY (PYY), Secretin, and Tumor Necrosis Factor alpha (TNFα). Notes: GLP-1 (Active) and GLP-1 (Total) cannot be run together in the same assay. Protease inhibitors should be added upon collection of samples.

Research Category: Metabolism & Endocrinology

Research Sub-Category:

  • Metabolic Syndrome
  • Obesity
  • Diabetes

Application

The MILLIPLEX® Rat Metabolic Hormone Expanded Panel is a bead-based quantitative assay based on the Luminex® xMAP® technology, which enables the simultaneous analysis of up to 14 analytes in a 25 µl sample volume. This is an overnight assay requiring 25 µL rat serum, plasma, or cell/tissue culture supernatants or lysates. No sample dilution is required.

Analytes Available: Amylin (active);C-Peptide 2; Ghrelin (active);GIP (total);GLP-1 (active);GLP-1 (total);Glucagon;IL-6;Insulin;Leptin;MCP-1/CCL2;Pancreatic Polypeptide (PP);PYY (total);Secretin;TNFα.

Cross Reactivity: There was no or negligible cross-reactivity between the antibodies and any of the other analytes in this panel.

Sensitivity: Refer to kit protocol for sensitivities of each analyte.

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

96 well plate

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results and see how these multiplex biomarker Luminex® assays are being used to advance research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service