RDYSTRM70
ReadyStream® RJ45 Cable
For use with ReadyStream® system to connect the dispensing unit and the media unit
Synonym(s):
Connection Cable
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
13102014
NACRES:
NB.51
Recommended Products
packaging
pkg of 1 unit
parameter
0.087 kg operating weight
cable L × W × H
20 cm × 20 cm × 3 cm
application(s)
food and beverages
compatibility
for use with ReadyStream®
General description
ReadyStream® RJ45 Cable is an accessory of the ReadyStream® instrument to connect the dispensing unit and the media unit.ReadyStream®system, with its media preparation and dispensing units, media bags, and other accessories, is a first-of-its-kind solution that prepares and instantly dispenses culture media for use in microbiological food testing.
Application
ReadyStream® RJ45 Cable is used for the connection between Media and Dispensing units.
Other Notes
To access the ReadyStream® Data Matrix Generator Tool follow this link https://www.sigmaaldrich.com/US/en/support/calculators-and-apps/readystream-data-matrix-generator
Legal Information
READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service