Skip to Content
Merck
All Photos(3)

Key Documents

RDYSTPC01

Millipore

Autoclavable 3-Way Stopcock

To control the media flow direction in the ReadyStream® workflow

Synonym(s):

3 way valve, valve connector

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€258.00

€258.00


Available to ship on11 April 2025Details



Select a Size

Change View
1 EACH
€258.00

About This Item

UNSPSC Code:
13102008
NACRES:
NB.51

€258.00


Available to ship on11 April 2025Details


material

PTFE

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 pieces

application(s)

food and beverages

General description

The autoclavable stopcock can optionally be connected between ReadyStream® System, GL45 lab bottle and dilutor to control the media flow direction

Legal Information

READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service