Skip to Content
Merck
All Photos(4)

Key Documents

PYR0MATKIT

Millipore

PyroMAT® kit

Reagents and Interleukin-6 ELISA kit for Monocyte Activation Test (MAT), for Pyrogen testing

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€966.00

€966.00


Estimated to ship on11 June 2025Details


Request more information

Select a Size

Change View
1 EA
€966.00

About This Item

UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NA.74

€966.00


Estimated to ship on11 June 2025Details


Request more information

Agency

EP Chapter 2.6.30

Quality Level

feature

ready-to-use

packaging

pkg of 1 kit

manufacturer/tradename

PyroMAT®

application(s)

pharmaceutical
pyrogen testing

compatibility

configured for PyroMAT®

shipped in

ambient

storage temp.

2-8°C

suitability

pyrogenic bacteria

General description

The PYROMAT® System is a ready-to-use, in vitro solution for pyrogen testing, using the Monocyte Activation Test (MAT). It is based on the Mono-Mac-6 cell line and interleukin 6 (IL-6) read-out. The human monocytes that encounter pyrogens from contaminated sample, produce cytokines (IL-6), which can be detected with an enzyme-linked immunosorbent assay (ELISA). The kit contains the reagents needed to prepare the sample, perform the incubation with the monocytic cells, and the reagents needed for the ELISA detection.

Application

The PYROMAT® System is used to ensure the safety of parenteral pharmaceutical products and medical devices.

Features and Benefits

Ready-to-use kit

Components

  • 96-well plate, Endotoxin free water, RMPI culture medium
  • IL-6 ELISA kit and plate

Storage and Stability

2-8°C

Other Notes

Download the PyroMAT® Method Software from here

Recommended products

To be used with PYROMAT® cells (PYR0MATCELLS), Reference standard endotoxin (144.161.0001), NEP controls (MATHKSA, MATFLAGELLIN) and IL6 control (PYR0MATIL6)

Legal Information

PYROMAT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service