Skip to Content
Merck
All Photos(1)

Key Documents

PX0511

Supelco

Phenol Liquefied

GR

Synonym(s):

Phenol solution, Hydroxybenzene, Carbolic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O
CAS Number:
Molecular Weight:
94.11
Beilstein:
6428463
MDL number:
UNSPSC Code:
12352104
Pricing and availability is not currently available.

Quality Level

Assay

≥88.0% (iodometric)

form

liquid

impurities

≤12% Water (H2O)

non-volatile residue (NVR)

≤0.05%

color

APHA: ≤50

transition temp

flash point 81 °C (Information refers to the main component.)

density

1.07 g/cm3 at 25 °C

shipped in

ambient

storage temp.

room temp

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Application

GR

Analysis Note

Assay (iodometric): 88.0% min
Color (APHA): 50 max
Congealing point: 10.5-14.0°C
Form: Clear liquid
Nonvolatile matter: 0.05% max
Water (H2O): 12% max

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup - Information refers to the main ingredient.

Flash Point(C)

81 °C - closed cup - Information refers to the main ingredient.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service