Skip to Content
Merck
All Photos(1)

Key Documents

PX0424

Supelco

Petroleum Ether Non UV

OmniSolv®

Synonym(s):

Petroleum ether, Ligroine,Petroleum Spirit, Benzine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105
bp:
30-50 °C/1013 hPa
Pricing and availability is not currently available.

Quality Level

product line

OmniSolv®

form

liquid

impurities

≤0.01% Water (H2O)
≤0.2 μeq/g Titrable acid
≤2 ng/L Capillary ECD responsive substances (as PCNB)

evapn. residue

≤1 ppm

color

APHA: ≤10

bp

30-50 °C/1013 hPa

transition temp

flash point <-30 °C

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Not Optically Transparent in UV
OmniSolv®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Boiling range30-60°CCapillary ECD responsive substances (as PCNB)2ng/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardTitrable acid0.2µeq/g maxWater (H2O)0.01% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 1 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

<-22.0 °F

Flash Point(C)

< -30 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service