Skip to Content
Merck
All Photos(1)

Key Documents

PSRP004

Millipore

Millicell® 96 well Plate

pore size 1 μm, PET membrane, sterile

Synonym(s):

96-Well Plate

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€507.00
5 EA
€2,220.00

€507.00


Estimated to ship on15 April 2025Details



Select a Size

Change View
1 EA
€507.00
5 EA
€2,220.00

About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.13

€507.00


Estimated to ship on15 April 2025Details


material

PET membrane
flat wells
polystyrene

Quality Level

description

Polyethylene terephthalate, 1 μm pore size, sterile, 96-well cell culture plate, single-well feeder tray, 96-well receiver tray and lid

sterility

sterile

feature

lid

manufacturer/tradename

Millicell®

parameter

50 °C max. temp.

technique(s)

cell attachment: suitable
cell culture | mammalian: suitable
cell differentiation: suitable
immunocytochemistry: suitable

H

26.2 mm

L

128 mm

W

84.8 mm

filter diam.

0.4 mm

filtration area

0.11 cm2

plate size

96 wells

working volume

75 μL

pore size

1 μm pore size

input

sample type hematopoietic stem cell(s)
sample type: human embryonic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type mesenchymal stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type neural stem cell(s)
sample type epithelial cells
sample type cardiac stem cell(s)
sample type pancreatic stem cell(s)

binding type

Tissue Culture (TC)-treated surface

shipped in

ambient

Related Categories

General description

Device Configuration: 96-well plate
Filter Code: PET
Millicell cell culture insert plates are a patented device designed to support cell growth and differentiation of endothelial and epithelial cell lines, including CaCo-2.

The Millicell device is available with a 0.4 µm PCF and a variety of PET membrane pore sizes. The plate is designed for maximum user convenience and incorporates features such as apical assist for easier pipetting and basolateral access. To reduce the risk of monolayer contamination, the membrane plates have feet to elevate the plate above the work surface when disassembled from the feeder tray. Tear drop-shaped receiver wells eliminate air bubbles as plates are assembled.

Millicell plate are optimized to grow and sustain high integrity cell monolayers. Cells grown on Millicell plate membranes grow better than on plastic because the cells are nourished from both the apical and basolateral sides. Cell growth and function more closely mimic in vivo. Millicell plates are designed for analysis as well as cell growth and can be used manually or with automated cell seeding, feeding and washing systems

With this complete system, you can grow, feed, and analyze cells in one membrane-bottom plate. The plate can be used with either a single-well or a multi-well feeding tray. At the time of transport analysis, the plate is simply transferred to a multi-well transport tray for analysis.

This streamlined design enhances functionality with:

- Seed-and-feed systems

- Automated liquid handling systems and span pipettes

- Basolateral and apical access to cells

- Transepithelial electrical resistance (TEER) measurement systems

Application

Research Category
Cell Culture

Packaging

Bulk packed

Components

96-well cell culture plate, single-well feeder tray, lid

Other Notes

Automation Compatible: Yes
5-pack excludes single-well trays product PSSW010R5.

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page covers the basic indirect co-culture procedure on both sides of Millicell cell culture insert membranes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service