Skip to Content
Merck
All Photos(1)

Documents

PRCYMAG40PMX23BK

Millipore

MILLIPLEX® Non-Human Primate Cytokine Magnetic Bead Panel - Premixed 23 Plex Space Saver (Bulk) Packaging

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in non-human primate serum, plasma and cell culture samples.

Synonym(s):

Luminex® non-human primate cytokine immunoassay, Millipore non-human primate cytokine panel, Monkey cytokine multiplex kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

nonhuman primates

manufacturer/tradename

Milliplex®

assay range

linearity: 110.3%
(1:06)

linearity: 121.6%
(1:12)

linearity: 126.2%
(1:24)

standard curve range: 12.2-50,000 pg/mL
(IL-10, IL-18)

standard curve range: 2.4-10,000 pg/mL
standard curve range: 4.9-20,000 pg/mL
(IL-4)

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The immune system is an organized complex network of biological structures and processes that protect against disease. For example, cytokine and chemokines mediate interactions between cells directly, regulating target immune cell responses. These responses result in inflammation where the immune system attempts to eradicate foreign antigens and begin the healing process. Consequently, the inflammatory process plays a key protective role in immunity. In addition, cytokine and chemokine research is essential in understanding the immune system and its multi-faceted response to most antigens, as well as disease states such as autoimmune disease, allergic reactions, sepsis, and cancer.

The MILLIPLEX® Non-Human Primate Cytokine premixed panel is to be used for the simultaneous quantification of the following in tissue/cell lysate and culture supernatant
samples and serum or plasma samples: G-CSF, GM-CSF, IFNγ, IL-1β, IL-1ra, IL-2, IL-4, IL-5, IL-6, IL8, IL-10, IL-12/23(p40), IL-13, IL-15, IL-17A, MCP-1, MIP-1β, MIP-1α, sD40L, TGF-α, TNF-α, VEGF, and IL-18.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
The antibody pairs in the panel are specific only to the desired analyte and exhibit no or negligible cross-reactivity with other analytes in the panel
UPDATE >>
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: G-CSF, GM-CSF, IFN-γ, IL-1RA, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-8, IL-10, IL-12/23 (p40), IL-13, IL-15, IL-17, IL-18, MCP-1, MIP-1α, MIP-1β, sCD40L, TGF-α, TNF-α, VEGF
  • Recommended Sample type: Serum, plasma and cell culture
  • Recommended Sample dilution: Neat
  • Assay Run Time: Overnight or one day. For best results, an overnight incubation is recommended
  • Research Category: Inflammation & Immunology

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities for individual cytokines/chemokines
Standard Curve Range: Refer to QC analysis sheet for exact concentration (sCD40L, TNFα)

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

G-CSF, GM-CSF, IFN-γ, IL-1ra, IL-1ß, IL-2, IL-5, IL-6, IL-8, IL-12/23 (p40), IL-13, IL-15, IL-17, MCP-1, MIP-1α, MIP-1ß, TGF-α, VEGF
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiwen Zheng et al.
Cell reports, 39(8), 110864-110864 (2022-05-21)
The pathological and immune response of individuals with COVID-19 display different dynamics in lung and intestine. Here, we depict the single-cell transcriptional atlas of longitudinally collected lung and intestinal tissue samples from SARS-CoV-2-infected monkeys at 3 to 10 dpi. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service