Skip to Content
Merck
All Photos(2)

Documents

PR0GTLCS1

Milli-Q

Progard® TL1 Pretreatment Pack

Protects and maintains good performance of the reverse osmosis (RO) membrane

Synonym(s):

Progard® TL1-C Pretreatment Pack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

Quality Level

feature

removal of large particles, organic compounds and free chlorine from tap water

packaging

pkg of 1 unit

manufacturer/tradename

Progard®

compatibility

for use with Elix®
for use with Elix® Gulfstream
for use with Elix® Large
for use with RiOs Large
for use with RiOs

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Proper pretreatment is an essential step in producing the desired final product water quality. The choice of the pretreatment is made according to the flow rate of the feed water.
The Progard® TL1-C Pretreatment Pack pretreats tap water by removing large particles, organic compounds and free chlorine. This protects the reverse osmosis (RO) membrane against mineral scaling, organic fouling and chlorine oxidation.
Intended for use with larger water purification systems: Elix® 20/35/70/100 and RiOs 30/50/100/150/200.

Application

For optimal protection of the RO membrane and automatic RO maintenance.

Features and Benefits

  • Optimal protection of the water system.
  • Ensures system lifetime and performance are optimized.
  • Polyphosphate - specifically selected to avoid scaling without releasing ions.
  • Depth-filtration for particle removal.
  • ROProtect CR for automatic RO maintenance.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
PROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
RIOS is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service