Skip to Content
Merck
All Photos(1)

Key Documents

PF060

Millipore

TdT, Calf Thymus

Sign Into View Organizational & Contract Pricing

Select a Size

0.155 ML
€311.00

€311.00


Estimated to ship on29 April 2025Details



Select a Size

Change View
0.155 ML
€311.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.71

€311.00


Estimated to ship on29 April 2025Details


form

liquid

Quality Level

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

General description

Native TdT from calf thymus. Tdt is a DNA polymerase that catalyzes the addition of deoxynucleotides to the 3′ hydroxy terminus of either double of single stranded DNA in a template-independent manner.
The Calbiochem® Terminal deoxynucleotidyl Transferase (TdT) is a DNA polymerase isolated from calf thymus. TdT catalyzes the addition of deoxynucleotides to the 3′-hydroxyl termini of either double or single stranded DNA molecules in a template independent manner. Although TdT preferentially adds deoxynucleotides to 3′-extensions, tailing onto 5′ overhangs or blunt ended double strands of DNA can also be achieved. TdT can be used for labeling DNA fragments and vectors with homopolymer tails and/or with modified deoxynucleotides such as biotin-dNTPs, 32P-dNTPs, cordycepin-dNTPs, or ddNTP.

Application

DNA Labeling

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is the amount of enzyme required to transfer 1 nmol of dAMP from dATP to the 3ʹ-OH terminus of d(A)50 in 60 min at 37°C.

Physical form

775 Units in 50 mM potassium phosphate buffer, 1 mM B-mercaptoethanol, 50% glycerol, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Chang, L.M.S. and F.J. Bollum. 1986. Crit. Rev. Biochem.21, 27.
Deng, G. and R. Wu. 1983. Methods Enzymol. 100, 96.
Michelson, A.M. and S.H. Orkin. 1982. J. Biol. Chem.257, 14773.
Roychoudhury, R. and R. Wu. 1980. Methods Enzymol.65, 43.
Nelson, T. and D. Brutlag. 1979. Methods Enzymol. 68, 41.
Roychoudhury, R., et al. 1976. Nucleic Acids Res.3, 101.
Bollum, F.J. 1974. In The Enzymes, 3rd edition (ed. P.D. Boyer), Vol. 10, 145. Academic Press, New York.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service