Skip to Content
Merck
All Photos(1)

Key Documents

PC223L

Sigma-Aldrich

Anti-Neuropeptide Y Rabbit pAb

lyophilized, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€512.00

€512.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€512.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€512.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

contains

≤0.1% sodium azide as preservative

species reactivity

human, avian, amphibian, fish, feline, rat, newt

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPY(4852)

General description

Rabbit polyclonal antibody supplied as lyophilized undiluted serum. Recognizes neuropeptide Y.
Recognizes neuropeptide Y in cortex, caudate, putamen, and paraventricular nucleus.
This Anti-Neuropeptide Y Rabbit pAb is validated for use in Frozen Sections for the detection of Neuropeptide Y.

Immunogen

porcine neuropeptide Y

Application


Frozen Sections (1:1000-1:2000 for free-floating, PAP detection)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Reconstitute the lyophilized antibody with 100 µl sterile distilled H₂O. Be careful to reconstitute the entire contents of the vial; during shipment and handling portions of the lyophilized pellet may have become dislodged and may not be in the bottom of the vial. Dilute with sterile PBS or Tris buffer at dilutions no higher than 1:10. Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C. Avoid freeze/thaw cycles of solutions.

Analysis Note

Positive Control
Paraventricular nucleus, cortex, or caudate putamen

Other Notes

Antibody specificity was examined in the rat cortex, caudate putamen, and paraventricular nucleus. Staining is completely eliminated by pretreatment of antibody with neuropeptide Y (10-6M). Preabsorption with avian pancreatic polypeptide, β-endorphin, cholecystokinin, peptide YY, somatostatin, or vasoactive intestinal peptide resulted in no loss of immunostaining. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service