Skip to Content
Merck
All Photos(1)

Key Documents

OP17L

Sigma-Aldrich

Anti-c-Fos (Ab-1) Mouse mAb (2G9C3)

lyophilized, clone 2G9C3, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
€472.00

€472.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
200 μG
€472.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€472.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2G9C3, monoclonal

form

lyophilized

does not contain

preservative

species reactivity

rat, human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG2a

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FOS(2353)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with AG.8653X mouse myeloma cells. Recognizes the ~55 kDa c-fos and the ~62 kDa v-fos proteins.
Recognizes both the ~55 kDa c-Fos and ~62 kDa v-Fos proteins in HeLa cells. Does not cross-react with the ~39 kDa Jun protein.
This Anti-c-Fos (Ab-1) Mouse mAb (2G9C3) is validated for use in Immunoblotting, Immunocytochemistry, Paraffin Sections for the detection of c-Fos (Ab-1).

Immunogen

Human
a synthetic peptide (KVEQLSPEEEEKRRIRRERNKMAAA) corresponding to amino acids 128-152 of human c-Fos

Application

Immunoblotting (2.5-5 µg/ml)

Immunocytochemistry (2.5-5 µg/ml)

Paraffin Sections (not recommended)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from a volatile buffer, 200 µg BSA.

Reconstitution

Reconstitute in sterile PBS, PH 7.4, or sterile Tris-saline (20 mM Tris containing 150 mM NaCl), pH 7.4, to a final concentration of 200 µg/ml. To achieve complete solubilization, reconstitute at (4°C) with 0.1% sodium azide or aliqout and freeze (-20°C) for long-term storage. Note: sodium azide is cytotoxic.

Analysis Note

Positive Control
HeLa cells

Other Notes

Jensen, D.E., et al. 1991. Oncogene6, 1219.
Reiss, M., et al. 1990. Cancer Res.50, 6641.
RouscherIII, F.J., et al. 1988. Science240, 1010.
Sassone-Corsi, P., et al. 1988. Cell54, 553.
Giardina, S.L., et al. 1987. Anal Biochem. 161, 109.
Verma, I.M., et al. 1987. Cancer Res.49, 29.
Verma, I.M., et al. 1987. Cell51, 513.
Muller, R. 1986. Viochem. Biophys. Acta.823, 207.
Verma, P. 1986. Trends Genet.2, 93.
Greenberg, M., et al. 1984. Nature (London)311, 433.
Recognizes both cellular and viral forms of fos. Does not react with the ~39 kDa jun protein. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service