Skip to Content
Merck

Skip To

NM0022

NovaSeal Die Kits

3/8 in. ID x 5/8 in. OD

Synonym(s):

Cutting Die for Electrical Crimping Tool

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit

Available to ship TODAYfromMerck Life Science BV/SRL

€887.00

About This Item

eCl@ss:
32010310
UNSPSC Code:
41121815

€887.00


Available to ship TODAYDetails


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

non-sterile

product line

NovaSeal

compatibility

for use with NovaSeal pinch pipes - NM0019PK100


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
NM0041NM0039NM0040
compatibility

for use with NovaSeal pinch pipes - NM0019PK100

compatibility

for use with NovaSeal pinch pipes - NM0019PK100

compatibility

for use with NovaSeal pinch pipes - NM0001PK100

compatibility

for use with NovaSeal pinch pipes - NM0002PK100

product line

NovaSeal

product line

NovaSeal

product line

NovaSeal

product line

NovaSeal

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

Quality Level

100

Quality Level

400

Quality Level

400

Quality Level

400


Still not finding the right product?

Explore all of our products under NovaSeal Die Kits




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters