Skip to Content
Merck
All Photos(1)

Key Documents

NA19L

Sigma-Aldrich

Anti-Replication Protein A (Ab-3) Mouse mAb (RPA34-20)

Synonym(s):

Anti-RP-A

Sign Into View Organizational & Contract Pricing


About This Item

antibody form

purified antibody

Quality Level

clone

RPA34-20, monoclonal

form

lyophilized

species reactivity

yeast, human, mouse

isotype

IgG2a

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with NS1 mouse myeloma cells. Recognizes the complexed (phosphorylated) and the ~34 kDa unphosphorylated subunit of replication protein A.
Recognizes the complexed (phosphorylated) and unphosphorylated subunits of replication protein A in HeLa and U293 cells and colon carcinoma tissue.
The replication system of a cell contains several essential components among which are replication factor A (RFA, RPA, or HeLa cell SSB), PCNA, and replication factor C (RFC) as well as several other proteins. RPA, a multi-subunit protein containing a 70 kDa, 34 kDa, and 11 kDa protein, may play a critical regulatory role in replication based on its functioning during both the initiation and elongation stages of DNA replication. The protein is highly conserved, having been identified in a range of species from yeast (Saccharomyces cerevisiae) to humans. The primary function of RPA appears to be as an auxiliary protein for both DNA polymerase α and δ. RPA undergoes a cell cycle dependent phosphorylation, which is restricted to the p34 protein. In addition to existing as part of the multi-subunit structure, p34 has also been identified as being present in a free, unphosphorylated form. Recent data shows that RPA is restricted to the nucleus and is overexpressed in transformed cells as well as in several tumor cells.

Immunogen

Epitope: within the p34 subunit of replication protein A
Human
replication protein A purified from U293 cells

Application

Immunoblotting (5 μg/ml)
Immunofluorescence (2.5 μg/ml)
Immunoprecipitation (1 μg/reaction)
Paraffin Sections (2.5 μg/ml, heat pre-treatment required)

Physical form

Lyophilized from 20 mM ammonium bicarbonate solution, 100 μg BSA.

Reconstitution

We recommend resuspending the lyophilized antibody with sterile phosphate buffered saline (PBS), pH 7.4, or sterile 20 mM Tris-saline (20 mM Tris containing 0.15 M NaCl), pH 7.4, to yield a final concentration of 100 μg/ml; product will be more stable

Analysis Note

Positive Control
HeLa or U293 cells or colon carcinoma tissue

Other Notes

Replication protein A (Ab-3) will react with both the complexed (phosphorylated) form of the protein as well as the free (unphosphorylated) 34 kDa subunit. Replication protein A can be used as a marker for cell division. Antibody should be titrated for op

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Tsurimoto et al.
The EMBO journal, 8(12), 3883-3889 (1989-12-01)
DNA synthesis by two eukaryotic DNA polymerases, alpha and delta, was studied using a single-strand M13 DNA template primed at a unique site. In the presence of low amounts of either DNA polymerase alpha or delta, DNA synthesis was limited
C R Wobbe et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(7), 1834-1838 (1987-04-01)
Simian virus 40 (SV40) DNA replication dependent on the SV40 origin of replication and the SV40 large tumor (T) antigen has been reconstituted in vitro with purified protein components isolated from HeLa cells. In addition to SV40 T antigen, these
S J Brill et al.
Nature, 342(6245), 92-95 (1989-11-02)
Cell-free replication systems for simian virus 40 (SV40) DNA are taken to be a model for the replication of eukaryotic chromosomes, because only one viral protein is required to supplement the replication proteins provided by a human cell extract. To
S Din et al.
Genes & development, 4(6), 968-977 (1990-06-01)
Replication factor A (RF-A) is a multisubunit, cellular protein that functions with SV40 T antigen during the initiation stage of DNA replication at the SV40 origin. It also cooperates with other replication factors to stimulate the activity of both polymerases
Initiation of eukaryotic DNA replication in vitro.
B Stillman
Annual review of cell biology, 5, 197-245 (1989-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service