Skip to Content
Merck
All Photos(1)

Key Documents

MXRPAIRTU

Millipore

Silicone air tubes

Spare part for Milliflex® Rapid AutoSpray Station

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
€19.80

€19.80


Estimated to ship on04 March 2025Details


Request more information

Select a Size

Change View
2 EA
€19.80

About This Item

UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.17

€19.80


Estimated to ship on04 March 2025Details


Request more information

material

silicone

packaging

pkg of 2 units

manufacturer/tradename

Milliflex®

input

sample type pharmaceutical(s)

application(s)

food and beverages
industrial qc
pharmaceutical

compatibility

AutoSpray Station

shipped in

ambient

General description

The silicone air tubes are spare parts that allow the Milliflex® Rapid AutoSpray Station to be maintained and deliver optimal reagent spraying performances.

Application

The silicone air tubes are Milliflex® AutoSpray Station accessory used to maintain and deliver optimal reagent spraying performances.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service