Skip to Content
Merck
All Photos(1)

Key Documents

MXPSITU37

Millipore

Silicone tubing

Milliflex® Plus Pump, liquid discard; 10 meters

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€76.20

€76.20


Estimated to ship on16 June 2025Details


Request more information

Select a Size

Change View
1 EA
€76.20

About This Item

UNSPSC Code:
41116129
NACRES:
NB.17

€76.20


Estimated to ship on16 June 2025Details


Request more information

material

silicone

packaging

pkg of 1 unit

manufacturer/tradename

Milliflex®

input

sample type pharmaceutical(s)

application(s)

bioburden testing
pharmaceutical
water monitoring

compatibility

Milliflex® Plus Pump

shipped in

ambient

General description

Silicone tubing is used for discarded the filtered liquid for Milliflex® Plus Pump.

Application

Silicone tubing is an accessory of the Milliflex® Plus Pump used during filtration of the test samples for bioburden testing, and water monitoring.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service