Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Estimated to ship on21 September 2026fromMerck Life Science BV/SRL | €1,870.00 |
About This Item
Quality Segment
manufacturer/tradename
Milliflex®
input
sample type pharmaceutical(s)
application(s)
industrial qc
shipped in
ambient
General description
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description The Milliflex PLUS Pumps validation protocol is available to make validation faster and easier and ensures that your system′s performance remains compliant with its validated specifications. | description The Milliflex Quantum validation protocol is available to make validation faster and easier and ensures that your system′s performance remains compliant with its validated specifications. | description The Milliflex Quantum validation protocol is available to make validation faster and easier and ensures that your system′s performance remains compliant with its validated specifications. | description The MAS-100 Iso MH Sampler validation protocol is available to make validation faster and easier and ensures that your system′s performance remains compliant with its validated specifications. |
| input sample type pharmaceutical(s) | input sample type pharmaceutical(s) | input sample type pharmaceutical(s) | input sample type pharmaceutical(s) |
| manufacturer/tradename Milliflex® | manufacturer/tradename Milliflex® Quantum | manufacturer/tradename Milliflex® Quantum | manufacturer/tradename MAS-100® |
| application(s) industrial qc | application(s) industrial qc | application(s) industrial qc | application(s) industrial qc |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
Still not finding the right product?
Explore all of our products under Milliflex PLUS Pumps Validation Protocol
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

