Skip to Content
Merck
All Photos(1)

Key Documents

MSTSTW375

Millipore

MAST® SP Sterile Tube Weld Assembly 0.375

new

For use with MAST® Autosampling Solution SP200 Sample Pilots

Synonym(s):

STW, Sample Pilot Connection, Sterile Tube Weld

Sign Into View Organizational & Contract Pricing


About This Item

Quality Level

storage condition

room temperature

General description

The MAST® SP STW Assembly 0.375 is designed for establishing aseptic Sterile Tube Weld (STW) links between SP200 Sample Pilots and in-process fluid lines such as a permeate outlet line. To be utilized, the line it′s inserted into must consist of CFLEX tubing with a 5/8-inch OD and 3/8-inch ID.

Application

MAST® Autosampling Solution
Automated Sampling, Process Analytical Technologies (PAT), Bioprocessing

Features and Benefits

  • Easy to connect
  • User friendly
  • Single Use

Packaging

10 units per box

Preparation Note

Autoclaved by User following connection to Sample Pilot

Other Notes

To be replaced after every autoclave cycle

Legal Information

MAST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Assessing on-line automated sampling as part of a PAT approach to shorten bioprocess development times through rapid, data-driven decisions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service