Skip to Content
Merck
All Photos(1)

Key Documents

MR-019

Sigma-Aldrich

EmbryoMax® CZB Medium With Phenol Red

The EmbryoMax CZB Medium With Phenol Red is available in a 50 mL format and has been optimized and validated for Embryo Culture.

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€50.70

€50.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
€50.70

About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.71

€50.70


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

manufacturer/tradename

Specialty Media
EmbryoMax®

input

sample type: mouse embryo(s)

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Mouse embryo media and embryo validated reagents for transgenic mouse embryo culture

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service