Skip to Content
Merck
All Photos(1)

Key Documents

MPE010025

Millipore

Microcon® Centrifugal Filters

MWCO 10 kDa, polyethersulfone membrane (Biomax), sample volume 0.5 mL

Synonym(s):

Biomax polyethersulfone (PES) membrane centrifuge filter

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
€115.00

€115.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 EA
€115.00

About This Item

UNSPSC Code:
41104916
NACRES:
NB.24

€115.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

material

polyacetal base (membrane support)
polycarbonate device top
polyethersulfone membrane (Biomax)
polypropylene tube (filtrate/concentrate)
silicone O-ring (medical grade)

sterility

non-sterile

product line

Microcon®

feature

holdup volume> 10 μL

parameter

0.5 mL sample volume

L

4.5 cm (1.8 in.)

diam.

12.3 mm

filtration area

0.32 cm2

pore size

10 kDa MWCO

General description

Filter Type: Ultrafiltration

Simply and efficiently concentrate and desalt solutions of any macromolecule, using any centrifuge that can accept 1.5 mL tubes.

With the Biomax membrane, Microcon® filters offer:
  • Reverse spin to maximize recovery, even in the smallest samples
  • Convenient storage of filtrate or concentrated sample in standard microfuge tube
  • Concentration factors up to 100X

Sample applications:
  • Plant

Application

These 0.5 mL centrifugal filter enables the concentration of biomolecules with a NMWL of 10kDa. Microcon® centrifugal filters are ideal for concentration & desalting.

Features and Benefits

  • High recovery for small volumes with reverse spin (concentration factor less than 20X)
  • Biomax membrane
  • Fast processing

Physical form

Color Code: Purple

Legal Information

MICROCON is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service