Skip to Content
Merck
All Photos(2)

Key Documents

MHA000P20

Millipore

Orange Serum Liquid Media Ampoule

capacity 2 mL, ampoule for cultvating aciduric microorganisms (yeast, mold, Lactic Acid Bacteria), particularly those associated with spoilage of citrus products

Synonym(s):

Orange Serum broth, OSA

Sign Into View Organizational & Contract Pricing

Select a Size

50 X 2 ML
€161.00

€161.00


Estimated to ship on02 June 2025Details


Request more information

Select a Size

Change View
50 X 2 ML
€161.00

About This Item

UNSPSC Code:
41106226
eCl@ss:
32160801
NACRES:
NA.74

€161.00


Estimated to ship on02 June 2025Details


Request more information

Quality Level

sterility

sterile

form

liquid

feature

ready-to-use

packaging

pack of 50 tests

capacity

2 mL

filling volume

2 mL

color

clear medium orange

application(s)

food and beverages
microbiology

compatibility

for use with MF-method
for use with Microfil®
for use with MicropreSure®
for use with Milliflex®

shipped in

ambient

suitability

molds
yeasts

General description

Orange Serum liquid media is developed specifically for the isolation and enumeration of micro-organisms that are capable of surviving in fruit juices and citrus products.{2} Fruit juices are generally acidic, with pH values ranging from approximately 2.4 for lemon juice, to 4.2 for tomato juice. The low pH of these foods is selective for yeast, moulds and a few groups of aciduric bacteria. Ready-to-use Liquid ampoule media solve numerous problems in microbiology laboratories where time, equipment and trained personnel are often in short supply. They are available in convenient, single test 2 mL plastic ampoules, eliminating the manipulation of glass ampoules and avoiding the waste and spoilage associated with bulk packaged media. Yeast colonies appear as white, creamy and large colonies. Bacteria colonies will be smaller white and transparent colonies.
Ready-to-use media solve numerous problems in microbiology laboratories where time, equipment and trained personnel are often in short supply. Our media are available in convenient, single test 2 mL plastic ampoules, eliminating the manipulation of glass ampoules and avoiding the waste and spoilage associated with bulk packaged media. Manufacturing and testing of ampouled media is conducted in strict accordance with precise quality control procedures to ensure optimal growth and maximum shelf life. Reagent grade water is used to clean glassware and make media. Other factors that affect media stability and performance, such as finished product pH, entrained gases, light exposure and temperature are carefully analyzed and controlled during the manufacturing process.

Application

Orange Serum liquid media is used for cultivating aciduric microorganisms (yeast, mold, Lactic Acid Bacteria), particularly those associated with spoilage of citrus products.{3}

Specifications

Incubation Time: 24–32 hrs

Analysis Note

Target Microorganisms: Lactic acid bacteria, yeast and mold
Yeast colonies appear as white, creamy and large colonies. Bacteria colonies will be smaller white and transparent colonies.

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Compendium of methods for the microbiological examination of foods. 2001
American Public Health Association and others
American Public Health Association (1985)
MacFaddin JF
Media for Isolation-Cultivation-Identification-Maintenance of Medical Bacteria (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service