Merck
MDE3007FS1
Millipore
Millistak+® Pod Depth Filter
DE30, filtration area 0.77m², Process Scale
Select a Size
1 EACH
€576.00
€576.00
Estimated to ship on21 May 2025Details
Select a Size
Change View
1 EACH
€576.00
About This Item
UNSPSC Code:
23151806
€576.00
Estimated to ship on21 May 2025Details
Product Name
Millistak+® Pod Depth Filter, DE30 (media series), cellulose membrane (and inorganic filter aid), effective filtration area 0.77 m2
material
TPE (Thermo Plastic Elastomer)
TPE gasket (Thermo Plastic Elastomer)
cellulose membrane (and inorganic filter aid)
polypropylene housing (glass-filled)
Quality Level
Sterilization Compatibility
autoclavable compatible
effective filtration area
0.77 m2
fitting
seal (flat seal)
cartridge code
DE30 (media series)
General description
The Millistak+® depth filter media is offered in a scalable, disposable format providing greater flexibility because of its unique modular design. The Millistak+® MDE3007FS1 comes in a POD format.
This DE30 media series delivers optimal performance through a gradient density matri, ideal for primary or secondary clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Features and Benefits
  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces
Legal Information
MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Material Qualification Dossier
Free of charge - Sign in to download
Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.