Skip to Content
Merck
All Photos(1)

Key Documents

MAC139

Sigma-Aldrich

Anti-Inntag 6 (IT6) Antibody, clone R19/4-11/15

clone R19/4-11/15, from mouse

Synonym(s):

Major hevein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€330.00

€330.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€330.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€330.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

R19/4-11/15, monoclonal

species reactivity

Hevea brasiliensis

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

General description

Pro-hevein (UniProt: P02877; also known as Major hevein) is encoded by the HEV1 gene in Hevea brasiliensis (Para rubber tree). Pro-hevein is a N-acetyl-D-glucosamine / N-acetyl-D-neuraminic acid binding lectin that can inhibit fungal growth. It is known to cause allergic reaction in human. It contains one barwin domain and one chitin-binding type-1 domain. Inntag 6 IT6 is a small protein domain of Hevea brasiliensis that displays strong structural determinants and does not generate solubility issues in vivo and in vitro. Also, it is not shown to compromise cell fitness when overexpressed and has no detectable functional and localization effects when fused to a variety of target proteins. Unlike the monomeric HA, Flag and MYC tags, Inntag IT6 does not increase the intrinsic variability of critical size in budding yeast cells. Structure of the Inntag IT6 is based on a packed beta-sheet conformation.

Specificity

Clone R19/4-11/15 detecs Pro-hevein in HEK293 cells tranfected with GFP-IT6.

Immunogen

His-tagged recombinant peptide fragment corresponding to 43 amino acids from Hevea brasiliensis.

Application

Anti-Inntag 6 (IT6), clone R19/4-11/15 is a mouse monoclonal antibody that detects the Innocuous Inntag IT6 wherein the tag protein is a fragment of Hev b 6.02 of Hevea brasiliensis. It has been tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Secondary & Control Antibodies
Western Blotting Analysis: A representative lot detected Inntag 6 (IT6) in Western Blotting applications (Georgieva, M.V., et. al. (2015). Nat Methods. 12(10):955-8).

Immunoprecipitation Analysis: A representative lot detected Inntag 6 (IT6) in Immunoprecipitation applications (Georgieva, M.V., et. al. (2015). Nat Methods. 12(10):955-8).

Immunofluorescence Analysis: A representative lot detected Inntag 6 (IT6) in Immunofluorescence applications (Georgieva, M.V., et. al. (2015). Nat Methods. 12(10):955-8).

Quality

Evaluated by Western Blotting in transfected lysates from HEK293cells transfected with GFP-IT6.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Inntag 6 (IT6) in 10 µg lysate from HEK293 cells transfected with GFP-IT6.

Target description

~34.1 kDa observed; 21.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service