Skip to Content
Merck
All Photos(1)

Key Documents

MABX2014

Sigma-Aldrich

Anti-HCG (Alpha-subunit) Combo Antibody, (10mg) KC

from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€129.00

€129.00


Estimated to ship on27 May 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
€129.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€129.00


Estimated to ship on27 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

General description

Human chorionic gonadotropin (HcG) is 237 amino acid glycoprotein that is released by the pacenta after implantation of an embryo. HcG is commonly used in diagnostics to detect pregnancy and depending on the sensitivityof the test, elevated levels of HcG can be detected in the Urine or Serum from as early as 3 weeks.
ELISA and Lateral Flow Immunoassay.

Immunogen

HCG Alpha subunit

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service