Skip to Content
Merck
All Photos(2)

Key Documents

MABT851M

Sigma-Aldrich

Anti-Podoplanin Antibody, clone PMab-1

clone PMab-1, from rat

Synonym(s):

Aggrus, Glycoprotein 38, Gp38, OTS-8, PA2.26 antigen, T1-alpha, T1A, Transmembrane glycoprotein E11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€373.00

€373.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€373.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€373.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

PMab-1, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Pdpn(14726)

Related Categories

General description

Podoplanin (UniProt: Q62011; also known as Aggrus, Glycoprotein 38, Gp38, OTS-8, PA2.26 antigen, T1-alpha, T1A, Transmembrane glycoprotein E11) I encoded by the Pdpn (also known as Gp38, Ots8) gene (Gene ID: 14726) in murine species. Podoplanin serves as the endogenous ligand of C-type lectin-like receptor-2 (CLEC-2) and is highly expressed in various tumors and in some normal cells, such as lymphatic endothelial cells and podocytes. Podoplanin is detected at high levels in lung and brain, at lower levels in kidney, stomach, liver, spleen and esophagus, and not detected in skin and small intestine. It is expressed in epithelial cells of choroid plexus, ependyma, glomerulus and alveolus, in mesothelial cells and in endothelia of lymphatic vessels. It is also expressed in stromal cells of peripheral lymphoid tissue and thymic epithelial cells and is detected in carcinoma cell lines and cultured fibroblasts. High expression of Podoplanin is observed in colon carcinomas. . It may be involved in cell migration and/or actin cytoskeleton organization. Podoplanin is localized to actin-rich microvilli and plasma membrane projections, such as filopodia, lamellipodia, and ruffles. Extensively O-glycosylated. Contains sialic acid residues. O-glycosylation is necessary for platelet aggregation activity. Podoplanin levels are down-regulated by treatment with puromycin aminonucleoside. Mice lacking or with defective podoplanin die at birth of respiratory failure due to a low number of attenuated type I cells, narrow and irregular air spaces, and defective formation of alveolar saccules. Administration of PMab-1 antibody is shown to reduce lymphangiogenesis in the corneal suture and ear wound healing murine models. It suppresses thioglycollate-induced macrophages infiltration at the site of wound healing. (Ref.: Maruyama, Y et al (2014). Invest.Ophthal. Visual Sci. 55(8); 4813-4822).

Specificity

Clone PMab-1 detects murine Podoplanin. It targets an epitipe within 14 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal region of murine Podoplanin.

Application

Anti-Podoplanin, clone PMab-1, Cat. No. MABT851, is a rat monoclonal antibody that detects mouse Podoplanin and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Podoplanin in mouse lung tissue.

Immunohistochemistry Analysis: A representative lot detected Podoplanin in Immunohistochemistry applications (Kaji, C., et. al. (2012) Acta Histochem Cytochem. 45(4):227-37).

Flow Cytometry Analysis: A representative lot detected Podoplanin in Flow Cytometry applications (Oki, H., et. al. (2015). Monoclon Antib Immunodiagn Immunother. 34(6):396-403).

Western Blotting Analysis: A representative lot detected Podoplanin in Western Blotting applications (Oki, H., et. al. (2015). Monoclon Antib Immunodiagn Immunother. 34(6):396-403).

Immunocytochemistry Analysis: A representative lot detected Podoplanin in Immunocytochemistry applications (Kaji, C., et. al. (2012) Acta Histochem Cytochem. 45(4):227-37).
Research Category
Cell Structure

Quality

Evaluated by Western Blotting in mouse colon tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Podoplanin in 10 µg of mouse colon tissue lysate.

Target description

~42 kDa observed; 18.23 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service