Skip to Content
Merck
All Photos(2)

Key Documents

MABT495

Sigma-Aldrich

Anti-MYL3, clone 7C1 Antibody

ascites fluid, clone 7C1, from mouse

Synonym(s):

Myosin light chain 3, Cardiac myosin light chain 1, CMLC1, Myosin light chain 1, slow-twitch muscle B/ventricular isoform, MLC1SB, Ventricular/slow twitch myosin alkali light chain

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€306.00

€306.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€306.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€306.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

7C1, monoclonal

species reactivity

rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

rat ... Myl3(24585)

General description

Myosin light chain 3 (MYL3), also known as Cardiac myosin light chain 1 (CMLC1) or Myosin light chain 1 slow-twitch muscle B/ventricular isoform (MLC1SB) or Ventricular/slow twitch myosin alkali light chain (VLC1) functions as a regulatory light chain of myosin. Myosins are a large superfamily of motor proteins that move along actin filaments, while hydrolyzing ATP. Myosin is the major component of thick muscle filaments, and is a long asymmetric molecule containing a globular head and a long tail. The molecule consists of two heavy chains and four light chains. Activation of smooth and cardiac muscle primarily involves pathways which increase calcium and myosin phosphorylation resulting in contraction. Myosin light chain phosphatase acts to regulate muscle contraction by dephosphorylating activated myosin light chain. MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Human myosin light chain has clinical application as a cardiac marker. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy.

Immunogen

Purified recombinant fragment of human MYL3 expressed in E. Coli

Application

Detect MYL3 using this Anti-MYL3, clone 7C1 Antibody validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected MYL3 in human skeletal muscle and cardiac muscle tissues.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blotting in rat fetal heart tissue lysate.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected MYL3 in rat fetal heart tissue lysate.

Target description

~25 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service