Skip to Content
Merck
All Photos(3)

Key Documents

MABT388

Sigma-Aldrich

Anti-ADAM 17 Antibody, clone 16C11.1 | MABT388

clone 16C11.1, from mouse

Synonym(s):

Disintegrin and metalloproteinase domain-containing protein 17, ADAM 17, Snake venom-like protease, TNF-alpha convertase, TNF-alpha-converting enzyme, CD156b

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€336.00

€336.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€336.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€336.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

16C11.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ADAM17(6868)

General description

Disintegrin and metalloproteinase domain-containing protein 17 (EC 3.4.24.86; UniProt P78536; also known as ADAM 17, CD156b, Snake venom-like protease. TNF-alpha convertase, TNF-alpha-converting enzyme, Tumor necrosis factor alpha-converting enzyme) is encoded by the ADAM17 (also known as CD156B, CSVP, NISBD, NISBD1, TACE) gene (Gene ID 6868) in human. Originally described as a TNF–alpha converting enzyme, ADAM17 is known to process the cleavage of many transmembrane proteins, including membrane-anchored cytokines, cell adhesion molecules, receptors, ligands, and enzymes. ADAM17 was the first ′sheddase′ identified. ADAM17 activity is regulated by multiple post-translational modifications. It has three phosphorylation sites on its C-terminal cytoplasmic domain (T735, S791, and (S819). ERK1/2-mediated T735 phosphorylation is reported to regulate ADAM17 maturation and surface expression. ADAM 17 is initially translated in its prepro-form and subsequent cleavages of the signal (aa 1-17) and propeptide (18-214) sequences produces the 610 aa mature ADAM 17 (aa 215-824).

Specificity

Expected to react with both spliced isoforms.

Immunogen

GST-tagged recombinant protein corresponding to human ADAM 17.

Application

Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This mouse Monoclonal Anti-ADAM 17 Antibody, clone 16C11.1, Cat. No. MABT388 detects levels of ADAM17, and has been published and validated for use in Western Blotting, Immunohistochemistry (Paraffin).
Western Blotting Analysis: 5.0 µg/mL from a representative lot detected ADAM 17 in 10 µg of human acute myelogenous leukemia KG-1 cell lysate.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected ADAM 17 in human testis and colorectal cancer tissues.

Quality

Evaluated by Western Blotting in human acute myelogenous leukemia KG-1 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected ADAM 17 in 10 µg of human acute myelogenous leukemia KG-1 cell lysate.

Target description

~120 kDa observed. A lower band ~100 kDa is also observed when using a higher antibody concentration (5 µg/mL). Due to glycosylation, target band size appears larger than the calculated molecular weights (93.0 kDa/prepro-isoform A, 68.2 kDa/mature isoform A, 78.5kDa/prepro-isoform B, 63.7 kDa/mature isoform B). Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service