Skip to Content
Merck
All Photos(2)

Key Documents

MABT386

Sigma-Aldrich

Anti-Hu GBP-1/GBP-1 Antibody, clone 6C5.1

clone 6C5.1, from mouse

Synonym(s):

Interferon-induced guanylate-binding protein 1, GTP-binding protein 1, GBP-1, HuGBP-1, Guanine nucleotide-binding protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on22 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6C5.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GBP1(2633)

General description

mgbp-2/hGBP-1, or also known as Interferon-induced guanylate-binding protein 1, GTP-binding protein 1 (GBP-1/HuGBP-1), or Guanine nucleotide-binding protein 1, and encoded by the gene GBP1, is another enzyme important in innate defense. Among its functions, this enzyme hydrolyzes GTP to GMP and appears to promote oxidative killing and deliver antimicrobial peptides to autophagolysosomes, thus providing broad host protection against different pathogen classes. mgbp-2/hGBP-1 is localized to the cytoplasm but is also secreted in endothelial cells and localizes to the cytoplasmic side of the Golgi apparatus in expressing most cells. Interestingly, mgbp-2/hGBP-1is secreted by the endothelial cells in the CNS upon bacterial challenge even when not induced by gamma interferon. The enzyme is also induced by TNF-alpha and IL-1beta expression by immune cells and is considered a proinflammatory response factor.

Immunogen

GST-tagged recombinant protein corresponding to hu GBP-1/GBP-1.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected hu GBP-1/GBP-1 in human spleen tissues and rat brain tissue.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Hu GBP-1/GBP-1 Antibody, clone 6C5.1 is validated for use in western blotting & IHC (Paraffin) for the detection of Hu GBP-1/GBP-1.

Quality

Evaluated by Western Blotting in human spleen tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected hu GBP-1/GBP-1 in 400 µg of human spleen tissue lysate.

Target description

~68 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service