Skip to Content
Merck
All Photos(2)

Documents

MABT379

Sigma-Aldrich

Anti-Anoctamin-1/TMEM16A Antibody, clone 5F8.1

clone 5F8.1, from mouse

Synonym(s):

Anoctamin-1, Discovered on gastrointestinal stromal tumors protein 1, Oral cancer overexpressed protein 2, Transmembrane protein 16A, Tumor-amplified and overexpressed sequence 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F8.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ANO1(55107)

General description

Discovered on gastrointestinal stromal tumors protein 1 (Dog1), also known as Anoctamin-1/TMEM16A, is a novel multi-pass membrane protein belonging to the anoctamin family. Anoctamin-1/TMEM16A functions as a Ca2+-activated Cl- channel and is a required component of tracheal development. Expression of Anoctamin-1/TMEM16A is elevated in the gastrointestinal stromal tumors (GISTs), KIT signaling-driven mesenchymal tumors of the GI tract. It is also expressed at more moderate levels in epithelia from foregut, kidney, lung, pancreas and salivary glands. Recent studies suggest that it may serve a roll as a diagnostic marker in the classification of GISTs.

Specificity

This antibody recognizes isoforms 1 and 2 of Anoctamin-1 based upon immunogen design.

Immunogen

Epitope: N-terminal cytoplasmic domain
GST-tagged recombinant protein corresponding to the N-terminal cytoplasmic domain of human Anoctamin-1/TMEM16A.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Anoctamin-1/TMEM16A in human prostate cancer sections.
This Anti-Anoctamin-1/TMEM16A Antibody, clone 5F8.1 is validated for use in western blotting & IHC for the detection of Anoctamin-1/TMEM16A.

Quality

Evaluated by Western Blotting in PC-3 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Anoctamin-1/TMEM16A in 10 µg of PC-3 cell lysate.

Target description

~114 kDa and ~98 kDa observed. Three isoforms are known to exist at 114 kDa (isoform 1), 98 kDa (isoform 2), and 74 kDa (isoform 3).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service