Skip to Content
Merck
All Photos(1)

Key Documents

MABS31

Sigma-Aldrich

Anti-Calcyclin Antibody, clone CPTC-Calcyclin-2

clone CPTC-Calcyclin-2, from mouse

Synonym(s):

S100 calcium binding protein A6, S100 calcium binding protein A6 (calcyclin), protein S100-A6, calcyclin, S100 calcium-binding protein A6 (calcyclin), Growth factor-inducible protein 2A9, Calcyclin, Prolactin receptor-associated protein, S100 calcium-bin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€286.00

€286.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€286.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€286.00


Estimated to ship on16 April 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CPTC-Calcyclin-2, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... S100A6(6277)

General description

Calcyclin, also known as S100A6, is a calcium binding protein that is highly expressed in fibroblasts and epithelial cells. Inhibition of calcyclin by p53 is part of the mechanism to control cell proliferation. Calcyclin is associated with cancer and metastasis and plays a role in cell cycle regulation.

Immunogen

Full length recombinant protein corresponding to human Calcyclin.

Material obtained from the National Cancer Institute’s Clinical Proteomic Technologies for Cancer (CPTC) program.

Application

Anti-Calcyclin Antibody, clone CPTC-Calcyclin-2 detects level of Calcyclin & has been published & validated for use in WB, ELISA, IH.
Immunohistochemistry Analysis: A previous lot was used by an independent laboratory in IH. (Tara Hitke, Developmental Studies Hybridoma Bank, University of Iowa.)
Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling

Quality

Evaluated by Western Blot in human lung tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Calcyclin on 10 µg of human lung tissue lysate.

Target description

~10 kDa observed

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human lung tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service