Skip to Content
Merck
All Photos(1)

Key Documents

MABS2071

Sigma-Aldrich

Anti-SUMO-1 Antibody, clone 21C7

clone 21C7, from mouse

Synonym(s):

Small ubiquitin-related modifier 1, GAP-modifying protein 1, GMP1, SMT3 homolog 3, Sentrin, Ubiquitin-homology domain protein PIC1, Ubiquitin-like protein SMT3C, Smt3C, Ubiquitin-like protein UBL1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€153.00
100 μG
€401.00

€153.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€153.00
100 μG
€401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€153.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

21C7, monoclonal

species reactivity

chicken, human, mouse, zebrafish, rat, Xenopus, fish

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SUMO1(7341)

General description

Small ubiquitin-related modifier 1 (UniProt: P63165; also known as SUMO-1, GAP-modifying protein 1, GMP1, SMT3 homolog 3, Sentrin, Ubiquitin-homology domain protein PIC1, Ubiquitin-like protein SMT3C, Smt3C, Ubiquitin-like protein UBL1) is encoded by the SUMO1 (also known as SMT3C, SMT3H3, UBL1, OK/SW-cl.43) gene (Gene ID: 7341) in human. SUMO-1 is a ubiquitin-like protein that can be covalently attached to proteins as a monomer or a lysine-linked polymer. Covalent attachment via an isopeptide bond to its substrates requires prior activation by the E1 complex SAE1-SAE2 and linkage to the E2 enzyme UBE2I, and can be promoted by E3 ligases such as PIAS1-4, RANBP2 or CBX4. This post-translational modification on lysine residues of proteins plays a crucial role in many cellular processes, such as nuclear transport, DNA replication and repair, mitosis and signal transduction. SUMO-1 can also covalently attach to the voltage-gated potassium channel KCNB1; this modulates the gating characteristics of KCNB1. Polymeric SUMO-1 chains are shown to be susceptible to polyubiquitination, which functions as a signal for proteasomal degradation of modified proteins. Two isoforms of SUMO-1 have been reported that are produced by alternative splicing. Mutations in SUMO1 gene are known to cause non-syndromic orofacial cleft 10, which is a birth defect consisting of cleft lips with or without cleft palate.

Specificity

Clone 21C7 detects SUMO-1 in multiple species. It targets an epitope with in 11 amino acids from the internal region.

Immunogen

His-tagged full length human recombinant Small ubiquitin-related modifier 1 (SUMO-1).

Application

Anti-SUMO-1, clone 21C7, Cat. No. MABS2071, is a mouse monoclonal antibody that detects Small ubiquitin-related modifier 1 (SUMO-1) and has been tested for use in Immunocytochemistry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected SUMO-1 in Immunohistochemistry applications (Rogers, R.S., et. al. (2004). Chromosoma. 113(5):233-43; Costa, M.W., et. al. (2011). PLoA One. 6(9):e24812).

Immunocytochemistry Analysis: A representative lot detected SUMO-1 in Immunocytochemistry applications (Matunis, M.J., et. al. (1996). J Cell Biol. 135(6 Pt 1):1457-70; Zhang, X.D., et. al. (2008). Mol Cell. 29(6):729-41).

Immunoprecipitation Analysis: A representative lot immunopprecipitated SUMO-1 in Immunoprecipitation applications (Costa, M.W., et. al. (2011). PLoA One. 6(9):e24812; Becker, J., et. al. (2013). Nat Struct Mol Biol. 20(4):525-31; Girach, F., et. al. (2013). Cell Rep. 5(5):1294-301; Chanda, A., et. al. (2017). PLoS One. 12(5):e0177639).

Western Blotting Analysis: A representative lot detected SUMO-1 in Western Blotting applications (Zhang, X.D., et. al. (2008). Mol Cell. 29(6):729-41; Matunis, M.J., et. al. (1996). J Cell Biol. 135(6 Pt 1):1457-70; Rogers, R.S., et. al. (2004). Chromosoma. 113(5):233-43; Costa, M.W., et. al. (2011). PLoA One. 6(9):e24812; Becker, J., et. al. (2013). Nat Struct Mol Biol. 20(4):525-31).

Immunofluorescence Analysis: A representative lot detected SUMO-1 in Immunofluorescence applications (Girach, F., et. al. (2013). Cell Rep. 5(5):1294-301; Rogers, R.S., et. al. (2004). Chromosoma. 113(5):233-43; Matunis, M.J., et. al. (1996). J Cell Biol. 135(6 Pt 1):1457-70).
Research Category
Signaling

Quality

Evaluated by Western Blotting in HCT116 cell lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected SUMO-1 in HCT116 cell lysate.

Target description

~17 kDa observed; 11.56 kDa calculated. SUMO-1/RanGAP1 band is observed AT ~ 75 kDa Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service