Skip to Content
Merck
All Photos(1)

Key Documents

MABS1989

Sigma-Aldrich

Anti-Osteocalcin Antibody, clone 4C5

clone 4C5, from mouse

Synonym(s):

Bone Gla protein, BGP, Gamma-carboxyglutamic acid-containing protein

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€130.00
200 μL
€353.00

€130.00


Estimated to ship on28 May 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
€130.00
200 μL
€353.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on28 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

4C5, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

dot blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... BGLAP(632)

Related Categories

General description

Osteocalcin (UniProt: P02818; also known as Bone Gla protein, BGP, Gamma-carboxyglutamic acid-containing protein) is encoded by the BGLAP gene (Gene ID: 632) in human. Osteocalcin is a member of the osteocalcin/matrix Gla protein family and can constitute 1 to 2% of the total bone protein. It is synthesized by osteoblasts and is a major component of the noncollagenous bone matrix. Osteocalcin is synthesized with a signal peptide (aa 1-23), a propetide (aa 24-51) that are cleaved to generate active secreted form of osteocalcin (aa 52-100). Osteocalcin displays a high affinity for calcium and binds strongly to apatite. Sixty to ninety percent of de novo synthesized osteocalcin is incorporated into the bone matrix where it binds to hydroxyapatite during matrix mineralization. The remainder of the osteocalcin is released into the circulation where it can be measured as a sensitive marker of bone formation. Osteocalcin found in serum is almost exclusively derived from the bone formation with little or no contribution from the resorption process. A major feature of osteocalcin is the presence of 3 vitamin K-dependent gamma-carboxyglutamic acid (Gla) residues, which are found in the first helical region and interact with the inter-calcium spacing in the HAP lattice. In humans, the percentage of the circulating osteocalcin that is not gamma-carboxylated is used as a biomarker of vitamin K status. Osteocalcin expression in osteoblasts is stimulated by insulin and adiponectin and osteocalcin enhances insulin sensitivity and differentiation in muscle. Serum osteocalcin is elevated in diseases characterized by increased bone turnover such as osteoporosis, hyperparathyroidism and Paget′s disease, and low in conditions associated with low bone turnover such as hypoparathyroidism and growth hormone deficiency. (Ref.: Kanazawa I (2015). World J Diabetes. 6(18): 1345 1354; Gundberg, CM et al. (2012). Adv Nutr 3 (2): 149-157).

Specificity

Clone 4C5 detects all forms of human osteocalcin.

Application

Anti-Osteocalcin, clone 4C5, Cat. No. MABS1989, is a moue monocloanl antibody that detects osteocalcin and has been tested for use in Dot Blot.
Dot Blot Analysis: A representative lot detected 3xGlu, Glu17, and 3XGla peptides for human OCN (Courtesy of Mathieu Ferron, Ph.D., Montreal Clinical Research Institute, Montreal, Canada).

Quality

Dot Blot Analysis: A 1:250 dilution from a representative lot detected Osteocalcin at a wide range of concentrations (0.8 to 1600 ng).

Target description

10.96 kDa calculated.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service