Skip to Content
Merck
All Photos(2)

Key Documents

MABS1976

Sigma-Aldrich

Anti-Del-1 Antibody, clone D345

clone D345, from mouse

Synonym(s):

EGF-like repeat and discoidin I-like domain-containing protein 3, Developmentally-regulated endothelial cell locus 1 protein, Integrin-binding protein DEL1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€130.00
100 μG
€354.00

€130.00


Estimated to ship on07 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€130.00
100 μG
€354.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on07 March 2025Details


Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

D345, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable (paraffin)
inhibition assay: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... EDIL3(10085)

General description

EGF-like repeat and discoidin I-like domain-containing protein 3 (UniProt: O43854; also known as Developmentally-regulated endothelial cell locus 1 protein, Integrin-binding protein DEL1, Del-1) is encoded by the EDIL3 (also known as DEL1) gene (Gene ID: 10085) in human. Del-1 is a calcium-dependent glycoprotein that is secreted by endothelial cells and is shown to be expressed in a developmentally restricted pattern during embryogenesis. In adult tissues it is expressed in endothelial cells and in macrophages. Del-1 is reported to prevent apoptosis in endothelial cell subjected to TNF alpha, etoposide, and anoikis treatment. This anti-apoptotic effect is shown to be mediated specifically through binding of integrin alphav beta3 by the RGD motif. Del-1 expression is reported to be inversely related to that of IL-17 that is shown to inhibit Del-1 via a GSK-3beta and C/EBPb-dependent mechanism. This inhibitory action can be reversed at the PI3K/Akt signaling induced by D-resolvins. It serves as an endothelial cell-secreted homeostatic factor that regulates local inflammation in several tissues. It is reported to interfere with beta2-integrin-dependent inflammatory cell adhesion to endothelium and restricts transendothelial migration of neutrophils. Del-1 is synthesized with a signal peptide (aa 1-23), which is subsequently cleaved off in the mature form. It has three EGF-like domains in its N-terminal region. EGF-like domains 2 and 3 form a rigid rod via an interdomain calcium ion binding site, while the long linker between EGF1 and EGF2 lends considerable flexibility to EGF1. Del-1 also has discoidin I-like domains at the C-terminal region that mediate its binding to phosphatidylserine. Two isoforms of Del-1 have been described that are produced by alternative splicing. (Ref.: Dasgupta, SK et al. (2012). Circulation 125 (13); 1664-1672; Wang, Z., et al. (2012). Surgery 151(2); 296-305; Maekawa, T et al. (2015). Nat. Commun. DOI: 10.1038/ncomms9272).

Specificity

Clone D345 detects human Developmentally-regulated endothelial cell locus 1 protein (Del-1).

Immunogen

GST-tagged full length human recombinant Developmentally-regulated endothelial cell locus 1 protein (Del-1).

Application

Detects Del-1 using this Anti-Del-1, clone D345, Cat. No. MABS1976, is a mouse monoclonal antibody that detects Developmentally-regulated endothelial cell locus 1 protein (Del-1) and has been tested for use in ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Inhibition assay, & Western Blot.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Del-1 in human cerebral cortex tissue sections.

ELISA Analysis: A representative lot detected Del-1 in ELISA applications (Dasgupta, S.K., et. al. (2012). Ciculation. 125(13):1664-72; Maekawa, T., et. al. (2015). Nat Commun. 6:8272).

Inhibition Analysis: A representative lot inhibited the uptake of BODIPY-labeled platelet microparticles by endothelium. (Dasgupta, S.K., et. al. (2012). Ciculation. 125(13):1664-72).

Western Blotting Analysis: A representative lot detected Del-1 in Western Blotting applications (Dasgupta, S.K., et. al. (2012). Ciculation. 125(13):1664-72).

Flow Cytometry Analysis: A representative lot detected Del-1 in Flow Cytometry applications (Dasgupta, S.K., et. al. (2012). Ciculation. 125(13):1664-72).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry (Paraffin) in human pancreas tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected Del-1 in human pancreas tissue sections.

Target description

53.76 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-Feng Chen et al.
Bone & joint research, 12(12), 734-746 (2023-12-12)
Therapeutic agents that prevent chondrocyte loss, extracellular matrix (ECM) degradation, and osteoarthritis (OA) progression are required. The expression level of epidermal growth factor (EGF)-like repeats and discoidin I-like domains-containing protein 3 (EDIL3) in damaged human cartilage is significantly higher than

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service